missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 555-aha-dUTP, 1 mM in TE buffer

Product Code. 10654193
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10654193 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10654193 Supplier Invitrogen™ Supplier No. A32762
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For brighter conjugates

Labeled 5-aminohexylacrylamido-dUTP (aha-dUTP) nucleotides are modified at the C-5 position of uridine with a unique hexylacrylamide linker, which serves as a spacer between the nucleotide and the dye. This spacer reduces interactions between the nucleotide and the dye, resulting in brighter conjugates and increased hapten accessibility for secondary detection reagents. Labeled nucleotides can be used to generate labeled nucleic acid hybridization probes for many molecular biology and molecular cytogenetics applications, including multicolor techniques.

Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeled Nucleotide
Concentration 1 mM
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Alexa Fluor Dye
Modification AHA (5-aminohexylacrylamido)
Shipping Condition Wet Ice
Product Line Alexa Fluor
For Use With (Application) Molecular Biology
Quantity 50 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.