missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 647 ATP (Adenosine 5'-Triphosphate, Alexa Fluor™ 647 2'-(or-3')-O-(N-(2-Aminoethyl) Urethane), Hexa(Triethylammonium) Salt)

Product Code. 10768994
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10768994 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10768994 Supplier Invitrogen™ Supplier No. A22362
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bright far-red fluorescent dye

Alexa Fluor 647 adenosine 5'triphosphate (Alexa Fluor 647 ATP) is a mixed-isomer comprised of the Alexa Fluor 647 fluorophore, an exceptionally bright far-red fluorescent dye attached to the 2' or 3' position of the ribose ring via an aminoethylcarbamoyl linker.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type 647 ATP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye Alexa Fluor™ 647
Quantity 100 μL
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.