Learn More
Invitrogen™ APF (Hydroxyl Radical, Hypochlorite or Peroxynitrite Sensor)
Product Code. 10729423
Click to view available options
Quantity:
470 μL
Unit Size:
Each
Product Code. 10729423
Brand: Invitrogen™ A36003
Used together, these two ROS indicators can selectively detect hypochlorite anion
Aminophenyl fluorescein (APF) and hydroxyphenyl fluorescein (HPF, H36004), two reactive oxygen species (ROS) indicators developed by T. Nagano, offer greater specificity and stability than dichlorodihydrofluorescein diacetate (H2DCFDA, D399).
  • APF and HPF show limited non-selective reactivity and relatively high resistance to light-induced oxidation
  • Both of these new fluorescein derivatives are nonfluorescent until they react with hydroxyl radical or peroxynitrite anion
  • APF will also react with hypochlorite anion
  • Upon oxidation, both APF and HPF exhibit bright green fluorescence (excitation/ emission maxima approx. 490/515nm), making them compatible with any instrument that can detect fluorescein
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 470 μL
Product Type ROS Indicator