missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ 665/676 (Lipid Peroxidation Sensor)

Product Code. 11574187
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11574187 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11574187 Supplier Invitrogen™ Supplier No. B3932
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BODIPY 665/676 is a lipophilic probe used to detect lipid peroxidation. It exhibits a change in fluorescence after interaction with peroxyl radicals within cells.

BODIPY 665/676 is a lipophilic probe used to detect lipid peroxidation. It exhibits a change in fluorescence after interaction with peroxyl radicals within cells. We recommend dissolving BODIPY 665/676 in high-quality anhydrous DMSO to attain a stock concentration of 1 to 10 mM.

Lipid peroxidation is the oxidative degradation of cellular lipids by free radicals from reactive oxygen species, which can degrade lipids containing carbon-carbon double bonds such as phospholipids and polyunsaturated fatty acids. Oxidation of lipids leads to the generation of lipid peroxides and can cause damage to cell membranes, resulting in changes to signal transduction pathways and eventually leading to cell death. Lipid peroxidation is involved in apoptosis and one of the main contributors to ferroptosis, an iron-dependent, non-apoptotic form of cell death. Oxidative stress from lipid peroxidation plays a role in aging, as well as pathologies such as cancer, atherosclerosis, and neurodegenerative diseases.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Far-red
Quantity 5 mg
For Use With (Application) Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Shipping Condition Room Temperature
Target Function Lipid peroxidation
Detection Method Fluorescence
Excitation/Emission 665/676 nm
Indicator ROS
Product Line BODIPY
Product Type Lipid Sensor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.