missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BODIPY™ FL ATP-γ-S, Thioester (Adenosine 5'-O-(3-Thiotriphosphate), BODIPY™ FL Thioester, Sodium Salt)

Product Code. 10614963
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10614963 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10614963 Supplier Invitrogen™ Supplier No. A22184
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BODIPY FL fluorophore linked through the γ-thiol of ATP-γ

BODIPY FL ATP-γ-S binds to ATP-binding proteins. To create this fluorescent nucleotide, the BODIPY FL fluorophore was linked through the γ-thiol of ATP-γ-S. In addition to its potential use for binding studies, the BODIPY FL ATP-γ-S thioether is an important new substrate for Fhit, a member of the histidine triad superfamily of nucleotide-binding proteins.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type FL ATP-gamma-S Thioester
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye BODIPY™ FL
Quantity 50 μL
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.