missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrainStain™ Imaging Kit

Product Code. 10674243
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10674243 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10674243 Supplier Invitrogen™ Supplier No. B34650
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with standard histochemical methods such as immunohistochemistry

The BrainStain Imaging Kit enables three-color combinatorial labeling of myelin, neurons, and nuclei in brain cryosections in a single 20-minute staining step. The kit contains novel stains that can be used together in one staining solution or separately and replaces traditional methods that can take one to three days.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green, Red, Blue
Detection Method Fluorescence
Product Type Imaging Kit
Quantity 1 kit
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye NeuroTrace™ 530⁄615, DAPI, FluoroMyelin™ Green
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Nucleus, Myelin, Nissl Bodies
Product Line BrainStain

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.