missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrdUTP (5-Bromo-2'-Deoxyuridine 5'-Triphosphate), 10 mM in TE buffer

Product Code. 10318172
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10318172 25 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10318172 Supplier Invitrogen™ Supplier No. B21550
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Label apoptotic cells

BrdUTP can be used in conjunction with tetrminal deoxynucleotidyl transferase (TdT) to label apoptotic cells in the TUNEL assay. BrdUTP can be detected with anti-BrdU antibodies.

Apoptosis, Cell Analysis, Cell Proliferation, Cell Viability, Proliferation and Function, Chromatin Biology, DNA Fragmentation, RNAi, Epigenetics and Non-Coding RNA Research

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type BrdUTP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 10 mM
Label or Dye Other Label(s) or Dye(s)
Quantity 25 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.