missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Brefeldin A (from Penicillium brefeldianum)

Product Code. 11584197
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11584197 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11584197 Supplier Invitrogen™ Supplier No. B7450
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The fungal metabolite brefeldin A (BFA) has multiple targets in cells and has proven valuable for dissecting the cellular processes, including vesicle formation and kinesin distribution, involved in exporting newly synthesized proteins.

The fungal metabolite brefeldin A (BFA) has multiple targets in cells and has proven valuable for dissecting the cellular processes, including vesicle formation and kinesin distribution, involved in exporting newly synthesized proteins. We are pleased to offer the natural brefeldin A isolated from Penicillium brefeldianum. Exposing cells to BFA causes a distortion in intracellular protein traffic from the ER to the Golgi apparatus and the eventual loss of Golgi apparatus morphology; removal of BFA completely reverses these effects. BFA also alters the morphology of endosomes and lysosomes.

Dissolve in high-quality anhydrous DMSO or ethanol to prepare a stock concentration up to 1 mM.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Shipping Condition Room Temperature
Label Type Unlabeled
Molecular Formula C16H24O4
Physical Form Powder
Product Type Sphingolipid
Molecular Weight (g/mol) 280.36
compliance-icons
Product Identifier
  • Brefeldin A (from Penicillium brefeldianum)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: brefeldin A

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.