missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C12FDG (5-Dodecanoylaminofluorescein Di-β-D-Galactopyranoside)

Product Code. 11590276
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11590276 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11590276 Supplier Invitrogen™ Supplier No. D2893
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Produces a green fluorescence

The β-galactosidase substrate, C12FDG has been covalently modified to include a 12-carbon lipophilic moiety. Once inside the cell, the substrate is cleaved by β-galactosidase producing a fluorescent product that is well retained by the cells, probably by incorporation of the liphophilic tail within the cell membrane. C12FDG is the main component in the ImaGene™ Green kit.

Beta-Galactosidase Assays, Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Flow Cytometry, Flow Cytometry Antibodies and Secondary Detection, Glycosidase, Neuraminidase, and Related Enzyme Activity, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Quantity 5 mg
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Excitation/Emission 490⁄514
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.