missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ Cell Reaction Buffer Kit

Product Code. 10297994
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10297994 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10297994 Supplier Invitrogen™ Supplier No. C10269
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

1 kit containing sufficient material for 50 reactions based upon a 0.5mL reaction volume.

Clicks the two moieties together

The Click-iT Cell Buffer Kit provides researchers who have cells labeled with an azide or alkyne and the corresponding click detection reagent with the means to click these two moieties together. Each kit includes sufficient material to perform 50 reactions based upon a 0.5mL reaction volume for subsequent analysis by flow cytometry, fluorescence microscopy or high-throughput imaging (HCS).

  • Less hazardous

Cell Analysis, Cell Proliferation with Click Chemistry, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Reaction Buffer Kit
Quantity 1 Kit
Content And Storage 1 kit containing sufficient material for 50 reactions based upon a 0.5 mL reaction volume.
Store at 2-6°C.
For Use With (Equipment) EdU, Click-it™ AHA, Alexa Fluor™ azide, Alexa Fluor™ alkyne, Click-iT™ GlcNAz, Click-iT™
Shipping Condition Room Temperature
Product Line Click-iT, Molecular Probes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.