missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ Nascent RNA Capture Kit, for gene expression analysis
GREENER_CHOICE

Product Code. 10283848
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10283848 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10283848 Supplier Invitrogen™ Supplier No. C10365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Component A: 1 x 5mg 5-ethynyl uridine (EU); Component B: 1 x 1mL Click-iT EU buffer 2X concentrate; Component C: 1 x 340μg biotin azide (PEG4 carboxamide-6-azidohexanyl biotin); Component D: 1 x 200μL copper (II) sulfate (CuSO4) 25mM aqueous solution; Component E: 1 x 400mg Click-iT reaction buffer additive 1; Component F : 1 x 100μLl Click-iT reaction buffer additive 2; Component G: 2 x 1mL Click-iT RNA binding buffer (2X concentrate); Component H : 1 x 500μL Dynabeads™ MyOne™ Streptavidin T1; Component I: 1 x 35mL Click-iT reaction wash buffer 1; Component J: 1 x 35mLl Click-iT™ reaction wash buffer 2

Perfect for researchers in academia and industrial settings

The Click-iT Nascent RNA Capture Kit, uses proprietary Click-iT chemistry to capture with high efficiency and sensitivity newly minted RNAs. By the Click-iT approach studies on high resolution analysis of gene regulation can be performed along with RNA regulation, RNA stability, RNA synthesis, RNA decay and degradation, transcriptional regulation, delta/delta C(t), nuclear run on/run off and more. Ethylene uridine (EU) ribonucleotide homologs containing an alkyne reactive group are fed to cells or tissue. Once incorporated, cells are lysed and the RNA isolated. A biotin azide is “clicked on” and then strepavidin magnetic beads are used to capture the newly synthesized pool of RNA. The captured RNAs are then amplified and the resultant cDNAs are used on any number of post-capture methodologies. We have validated it for arrays (both high and low density), sequencing on SOLiD™ and either SYBR™ or TaqMan™ based qPCR based delta CT analysis. As few as 25,000 cells are needed for the analysis of newly induced transcripts. The kit allows for 40 conditions for labeling and capture that can then be divided into 400 separate analytical assays. With a price of around dollar an assy this is perfect for researchers in academia and industrial settings. The EU reagent at the recommend concentration of 200μM is well tolerated by cells. In an array screen of over 32,000 genes, only the most minor changes in 12 compared to a control vehicle could be detected. Furthermore at initial feeding concentration 5x of what we recommend (200μM vs 1.0mM) there were no effects on a panel of housekeeping genes, or on the cells ability to proliferate normally. Sensitivity and reliability was confirmed in a low density array (TLDA™) of apoptotic genes. The captured nascent pool accurately found all the previously characterized apoptotic (stauroporine induced) transcripts, without any detectable change in sequence information.

  • Less hazardous

Discovery not just recovery. The last decades Genomic efforts revealed that only 2% of the human encodes protein, leaving 98% of the human genome as the Unknome. Discovering the purpose and expression patterns of this vast reservoir of unknown sequence will be markedly aided in this decade's transcriptomics by this new approach.

Click Chemistries, DNA and RNA Purification and Analysis, Gene Expression Analysis and Genotyping, Labeling Chemistry, Microarray Analysis, Nascent RNA Detection and Capture with Click Chemistry, Nucleic Acid Labeling and Oligo Synthesis, RNA Isolation for Expression Microarrays, RNA Labeling

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Component A
1 X 5 mg 5-ethynyl uridine (EU)
Component B
1 X1 ml Click-iT™ EU buffer *2X concentrate*
Component C
1 X 340 μg biotin azide (PEG4 carboxamide-6-azidohexanyl biotin)
Component D
1 X 200 μl copper (II) sulfate (CuSO4) *25mM aqueous solution*
Component E
1 X 400 mg Click-iT™ reaction buffer additive 1
Component F
1 X 100 μl Click-iT™ reaction buffer additive 2
Component G
2 X 1 ml Click-iT™ RNA binding buffer *2X concentrate*
Component H
1 X 500 μl Dynabeads™ MyOne™ Streptavidin T1
Component I
1 X 35 ml Click-iT™ reaction wash buffer 1
Component J
1 X 35 ml Click-iT™ reaction wash buffer 2
Store at 4 degrees C
Concentration 2X (EU Buffer & RNA Binding Buffer)
Format Kit
For Use With (Equipment) 7000 System, 7200 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Fast Mode, StepOne™, Standard Mode, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Veriti Thermal Cycler, Agilent 2100 Bioanalyzer, SOLiD™
Isolation Technology Magnetic Bead
Product Line Click-iT
Product Type Nascent RNA Capture Kit
Quantity 1 kit
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.