missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAF-FM (4-Amino-5-Methylamino-2',7'-Difluorofluorescein)

Product Code. 10675183
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10675183 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10675183 Supplier Invitrogen™ Supplier No. D23841
  • ONLINE EXCLUSIVE PRICE
    1334.00 €
    1080.54 € / 1mg
    Save 253.46 €
    19% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use to detect and quantify low concentrations of nitric oxide

DAF-FM is essentially nonfluorescent until it reacts with NO to form fluorescent benzotriazole. DAF-FM fluorescence can be detected by any instrument that can detect fluorescein, including flow cytometers, microscopes, fluorescent microplate readers and fluorometers.

  • Ex/Em of DAF-FM: approx. 495/515nm
  • Lyophilized product should be dissolved using DMSO and then added to an aqueous buffer to create a working solution
  • DAF-FM can be introduced into cells by pressure injection or perfusion from patch-clamp pipette
  • Buffers containing bovine serum albumin (BSA) and phenol red may affect the fluorescence and should be used with caution
  • Fluorescence quantum yield of DAF-FM is ∽0.005, but increases about 160-fold, to ∽0.81, after reacting with NO
  • Assessment of NO production in transaldolase-deficient lymphoblasts by flow cytometry
  • Detection of NO accumulation in embryonic cortical neurons following neurotrophin stimulation
  • in vivo imaging of NO in zebrafish
  • Intravital microscopic detection of NO generation associated with angiogenesis in mice
  • Quantitation of ATP-induced NO release in rabbit platelets
  • Spectra of NO adduct of DAF-FM are independent of pH above pH 5.5
  • NO adduct of DAF-FM is significantly more photostable than that of DAF-2, which means additional time for image capture
  • DAF-FM is more sensitive reagent for NO than is DAF-2 (NO detection limit for DAF-FM approx. 3nM versus approx. 5nM for DAF-2)

Refer to the Molecular Probes™ Handbook for additional product information.

Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Nitric Oxide Research, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Detection Method Fluorescence
Product Type NO Indicator

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.