missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDAO galactoside (9H-(1,3-Dichloro-9,9-Dimethylacridin-2-One-7-yl) β-D-Galactopyranoside)

Product Code. 11540346
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11540346 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11540346 Supplier Invitrogen™ Supplier No. D6488
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

The galactosidase substrate, DDAO galactoside yields a hydrolysis product that can be excited with the 633 nm laser (excitation/emission maxima ∼645/660).

The galactosidase substrate, DDAO galactoside yields a hydrolysis product that can be excited with the 633 nm laser (excitation/emission maxima ∼645/660). Although the substrate itself is fluorescent (excitation/emission maxima ∼460/610 nm), the difference between the substrate's and the hydrolysis product is over 200 nm, along the two species to be easily distinguished.

To make a concentrated stock solution of up to 5 mg/mL, dissolve in high quality anhydrous DMSO. Store frozen, desiccated and protected from light.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
Quantity 5 mg
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Excitation/Emission 465/608 nm
Molecular Weight (g/mol) 470.31
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.