missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dihydroethidium (Hydroethidine)

Product Code. 10530533
Change view
Click to view available options
Quantity:
25 mg
10 x 1 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10530533 10 x 1 mg 10mg
11520196 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10530533 Supplier Invitrogen™ Supplier No. D11347
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Free radical sensor

  • Superoxide indicator dihydroethdium, also called hydroethidine, exhibits blue-fluorescence in cytosol until oxidized, where it intercalates within cell's DNA, staining its nucleus a bright fluorescent red
  • Excitation Emission: 518/606nm

Cat. No. D23107 packaged 5mM solution stabilized in DMSO.

Apoptosis, Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing, Reactive Oxygen Species

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Quantity 10 x 1 mg
Excitation/Emission 518/606 nm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.