missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dihydroethidium (Hydroethidine)
Free radical sensor
506.00 € - 570.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10530533
|
Invitrogen™
D11347 |
10 x 1 mg |
570.00 €
10mg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11520196
|
Invitrogen™
D1168 |
25 mg |
506.00 €
25mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Superoxide indicator dihydroethdium, also called hydroethidine, exhibits blue-fluorescence in cytosol until oxidized, where it intercalates within cell's DNA, staining its nucleus a bright fluorescent red
- Excitation Emission: 518/606nm
Cat. No. D23107 packaged 5mM solution stabilized in DMSO.
Apoptosis, Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
For Research Use Only. Not for use in diagnostic procedures.