missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dil Stain (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate ('DiI'; DiIC18(3)))

Product Code. 11580276
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11580276 100 mg 100mg
11520326 25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11580276 Supplier Invitrogen™ Supplier No. D282
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DiI is a lipophilic membrane stain that diffuses laterally to stain the entire cell. It is weakly fluorescent until incorporated into membranes.

DiI is a lipophilic membrane stain that diffuses laterally to stain the entire cell. It is weakly fluorescent until incorporated into membranes. This orange/red-fluorescent dye, which is spectrally similar to tetramethylrhodamine, is often used as a long-term tracer for neuronal and other cells. DiI is also available as a solution (Cat. No. V-22885), as a paste (Cat. No. N-22880) or as a solid (Cat. No. D-282).

Prepare stock solutions of lipophilic tracers in dimethyl formamide (DMF), dimethylsulfoxide (DMSO), or ethanol at 1 to 2.5 mg/mL.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red-Orange
Content And Storage Store at room temperature and protect from light.
Excitation Wavelength Range 549 nm
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Emission 565 nm
Shipping Condition Room Temperature
Product Type Liphophilic Tracer
Sub Cellular Localization Cell Membranes, Lipids
I'm labeling live cells with Vybrant DiI or DiD lipophilic cyanine dyes. DiI gives a nice even membrane labeling, but DiD is more "spotty". What can be done?

This is expected. DiD (which is far-red fluorescent) is never as uniform as DiI (which is orange fluorescent). If uniformity is desired, try increasing the label time and concentration, but it still isn't likely to be as uniform as DiI. CellMask Deep Red Plasma Membrane stain is much more uniform and is about the same wavelength as DiD. However, if you intend to do cell tracking over days, CellMask stain has not been tried for that application.

I want to perform a cell fusion assay, where one cell line is labeled with one color and the other cell line with another color, and combine with a nucleic acid stain. What do you recommend?

A typical method is to label one cell line with orange fluorescent DiI C18 and the other cell line with green fluorescent DiO C18. These orange and green lipophilic cyanine dyes will stain the membranes of cells. Cells that fuse will then have both dyes, yielding a yellow color (when images are overlaid or cells are imaged in a dual-bandpass filter). These live cells can then be labeled with Hoechst 33342 (a cell-permeant blue DNA stain comparable in wavelength to DAPI), but only as an endpoint just before imaging (since DNA stains can interrupt DNA function).

I need to look at live cell morphology deformation over the course of a few hours. What sort of membrane dye would be useful for this?

Lipophilic cyanine dyes, such as DiI (Cat. No. D282), DiO (Cat. No. D275), DiD (Cat. No. D7757) or DiR (Cat. No. D12731), are commonly used. The longer the alkyl chain on the dye, the better the retention in lipophilic environments.

Why do I lose all signal from my neuronal tracer when I do a methanol fixation on my cells?

If the tracer you chose is a lipophilic dye and fix with methanol, the lipids are lost with the methanol. If you have to use methanol fixation then choose a tracer that will covalently bind to proteins in the neurons.

I stained my cells with a lipophilic cyanine dye, like DiI, but the signal was lost when I tried to follow up with antibody labeling. Why?

Since these dyes insert into lipid membranes, any disruption of the membranes leads to loss of the dye. This includes permeabilization with detergents like Triton X-100 or organic solvents like methanol. Permeabilization is necessary for intracellular antibody labeling, leading to loss of the dye. Instead, a reactive dye such as CFDA SE should be used to allow for covalent attachment to cellular components, thus providing for better retention upon fixation and permeabilization.

I labeled my neurons with DiI and then fixed and permeabilized and now I have no signal. What did I do wrong?

DiI is a lipophilic dye that resides mostly in lipids in the cell, when cells are permeabilized with detergent or fixed using alcohol this strips away the lipid and the dye. If permeabilization is required CM-DiI can be used because this binds covalently to proteins in the membrane; some signal is lost upon fixation/permeabilization, but enough signal should be retained to make detection possible.

How long does it take for lipophlic tracers to transport along the membrane? How much faster are the FAST lipophilic dyes?

The transport is fairly slow, around 6 mm/day in live tissue and slower in fixed tissue, so diffusion of lipophilic carbocyanine tracers from the point of their application to the terminus of a neuron can take several days to weeks The FAST DiO and DiI analogs (which have unsaturated alkyl tails) can improve transport rate by around 50%.

Which form of the lipophilic tracers (DiO, DiI, DiD, etc) should I use?

Select the dye that is compatible with your available excitation source(s) and emission filter set/channels. The solid, paste and crystal forms can be applied directly to neurons in tissues. For labeling cells in culture or microinjection, the lipophilic dyes in solution or solid form can be used.

How do I know which tracer to choose for my experiment?

Factors to consider are size of tracer, method of delivery (injection, direct application to tissue, etc.), and if the tracer needs to be fixable. Here are some links to details about the various classes of neuronal tracers we offer and how to choose between them:

Neuronal Tracing (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html)
Choosing a Tracer (https://www.thermofisher.com/us/en/home/references/molecular-probes-the-handbook/fluorescent-tracers-of-cell-morphology-and-fluid-flow/choosing-a-tracer.html)
Imaging Analysis (http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/bioprobes-50-journal.pdf)

Is there a way to label individual neurons without microinjecting?

The solid and crystalline forms of DiI and other related dyes (Cat. Nos. D282, D3911, D7757, and D12731) are sometimes placed in contact with a specific neuron where it will travel down the cell by lateral diffusion via the membrane. Alternatively, our NeuroTrace Tissue Labeling Paste can be scooped onto a needle and placed onto particular neurons.

Please see the information below for a comparison of our neuronal cell labeling methods:
Product:Method of labeling: Labeling intensity: Features
Neuron-specific antibodies: Primary antibodies directed to proteins expressed in neuronal cells: Proportional to the amount of protein expressed: Provides the only neuronal specific labeling method
Lipophilic neuronal ytracers: Hydrophobic dyes are incorporated into lipids in the cell: This labeling method provides the most intense labeling becuase of the abundant amount of lipids: Allows tracing of neurons throughout the sample
Membrane potential indicators: Dyes are loaded into live cells in aqueous buffers: Depends on either changes in structures due to the electrical field they are in, or dye influx due to depolarization: Changes in membrane potential play a central role in physiological processes, including nerve-impulse propagation, muscle contraction, and cell signaling

What products do you have for neuronal tracing?

Please check out this web page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/neuronal-tracing.html) for details.

I want to label two cell populations and then perform a cell fusion assay. Which reagents are best for imaging this?

Lipophilic cyanine dyes are preferred for this sort of assay, since they insert into cellular membranes and then, upon fusion, are shared by the fused cells as the membranes are shared. For example, one cell population can be labeled with DiI (orange-red) and another cell population can be labeled with DiO (green), and when the cells fuse, the combined color appears yellow (when imaged with a dual-bandpass filter set).

I want to track my cells with a nucleic acid stain, like DAPI or Hoechst dye. Do you recommend this?

This is not recommended. When these stains bind to DNA and RNA, they may affect the normal function of the nucleic acids, disrupting transcription, as well as replication. Other reagents, such as CellTracker dyes or Qtracker reagents are more optimized for tracking without disrupting normal activity. If a nuclear label is still desired, though, and the cells are mammalian and non-hematopoietic, CellLight nuclear reagents can transiently transfect cells to express GFP or RFP on a nuclear-expressing protein for up to several days without affecting function.

I want to track my cells over time, and you have a lot of options to choose from. How do I pick the right one?

Please see this Web link (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/cell-tracking.html) to help you choose the right option for your application. Start by planning how long you want to track your cells, then consider the mechanism of binding. Calcein dyes are very uniform in label and are good for short-term cell migration, but may be rapidly effluxed from some cell types. Lipophilic cyanine dyes, such as DiI, DiO, and similar dyes label cell membranes, don’t disrupt function, and can last longer, but have the potential to cross to other cells if membranes fuse. They are also lost upon permeabilization. CellTracker dyes are better for longer-term labeling, as they possess a mildly reactive chloromethyl moiety that allows covalent binding to cellular components. CFDA SE also covalently binds to cellular components. With all the reagents, their retention within cells is dependent upon the rate of cell division and the inherent properties of the cell (active efflux, membrane and protein turnover rates, etc.) and reagents that allow for covalent attachment exhibit longer retention than those that do not.

The longest-lasting and brightest options are the Qtracker reagents, which are taken up through endocytosis. These are so bright individual quantum dots can be detected, and are also robust enough to survive not only fixation and permeabilization, but even the heat and solvents used in paraffin processing.

I want to label the plasma membrane of my cells, but there are several dyes to choose from. Which one should I use?

For live-cell imaging, the CellVue and CellMask Plasma Membrane Stains are the most uniform and the slowest to be endocytosed. However, they are not the best choice if you wish to fix and permeabilize your cells, such as for antibody labeling. Wheat germ agglutinin (WGA) conjugates are also able to label live cells, or can label already formaldehyde-fixed cells. They can survive subsequent permeabilization with detergents, such as Triton X-100. If cells are already permeabilized, WGA will label internal structures as well. Thus, only an antibody against a plasma membrane protein can be used if cells are already permeabilized. Lipophilic cyanine dyes, such as DiI, will label all cell membranes in live cells, not just plasma membranes, if left on live cells for extended periods. Following page will help you choose (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-structure/plasma-membrane.html).


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.