missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DiO'; DiOC18(3) (3,3'-Dioctadecyloxacarbocyanine Perchlorate)

Product Code. 11570276
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11570276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11570276 Supplier Invitrogen™ Supplier No. D275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The green fluorescent, lipophilic carbocyanine >DiOC18(3) is widely used as a lipophilic tracer. It is weakly fluorescent in water, but highly fluorescent and quite photostable when incorporated into membranes.

The green fluorescent, lipophilic carbocyanine DiOC18(3) is widely used as a lipophilic tracer. It is weakly fluorescent in water, but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (∼1 nanosecond) in lipid environments. Once applied to cells, the dye diffuses laterally within the plasma membrane.

We recommend preparing stock solutions of lipophilic tracers in dimethyl formamide (DMF), dimethylsulfoxide (DMSO), or ethanol at 1 to 2.5 mg/mL. DMF is preferable to ethanol as a solvent for DiO.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store at room temperature and protect from light.
Excitation Wavelength Range 484 nm
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Emission 501 nm
Shipping Condition Room Temperature
Product Type Liphophilic Tracer
SubCellular Localization Cell Membranes, Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.