Learn More
Invitrogen™ DQ™ Gelatin From Pig Skin, Fluorescein Conjugate - Special Packaging
Description
Our DQ gelatin is a fluorogenic substrate that can be used to detect protease activity in vitro with high sensitivity. This substrate consists of highly quenched, fluorescein-labeled gelatin. Upon proteolytic digestion, its bright, green fluorescence is revealed and can be used to measure enzymatic activity. The fluorescence increase can be monitored with a fluorescence microplate reader or fluorometer.
Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Form | Powder |
| Product Line | DQ |
| Quantity | 5 x 1 mg |
| Substrate | Protease Substrate |
| Substrate Properties | Protein-Based Substrate |
| Target Enzyme | Metalloproteinase |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.