missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Cellulase Substrate, blue fluorescent, 339/452

Product Code. 10595403
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10595403 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10595403 Supplier Invitrogen™ Supplier No. E33953

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mixed with enzyme to produce fluorescent reaction

The EnzChek cellulase substrate was developed for simple and rapid quantitation of cellulase. The assay can be completed in 30 minutes or less. Simply mix the enzyme with the EnxChekcellulase substrate working solution, incubate, and read the results using excitation/emission maxima approx. 339/452nm. In contrast to other more complex, multistep assays, the EnzChekcellulase substrate is perfect for use in a high-throughput environment. This fluorescence substrate is highly sensitive, with a detection limit as low as 40μU/mL cellulase. It is also possible to use this substrate in a colorimetric assay, albeit with reduced sensitivity.

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, High-Content Screening (HCS), Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Colorimetric, Fluorescence
Form Solid
Product Line EnzChek
Quantity 1 mg
Substrate Cellulase substrate
Substrate Properties Chemical Substrate
For Use With (Equipment) Fluorometer, Microplate Reader
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.