missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoReporter™ lacZ/Galactosidase Quantitation Kit

Product Code. 10534803
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10534803 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10534803 Supplier Invitrogen™ Supplier No. F2905
  • ONLINE EXCLUSIVE PRICE
    497.00 €
    402.57 € / Pack of 1
    Save 94.43 €
    19% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reaction produces blue fluorescent product

The FluoReporter™ lacZ/Galactosidase Quantitation Kit includes the β-galactosidase substrate, 3-carboxyumbelliferyl β-D-galactopyranoside that produces the blue-fluorescent hydrolysis product, 7-hydroxycoumarin-3-carboxlic acid.

Beta-Galactosidase Assays, Cell Analysis, Enzyme and Protein Activity Assays, Flow Cytometry, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Beta-Galactosidase Reporter Gene Assay System
Detection Method Fluorescence
Assay β-Gal (lacZ) Assay
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Substrate CUG
Substrate Type Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Format Kit
Product Line FluoReporter
Quantity 1 Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.