Learn More
Invitrogen™ Fluorescein Di-β-D-Galactopyranoside (FDG)
Description
Cell permeant
Fluorescein di-β-D-galactopyranoside (FDG) is one of the most sensitive substrates for galactosidases. Nonfluorescent FDG is sequentially hydrolyzed by β-galactosidase, first to fluorescein monogalactoside (FMG) and then to highly fluorescent fluorescein. Enzyme-mediated hydolysis of FDG can be followed by the increase in either absorbance or fluorescence. Sensitive substrate for galactosides.
Beta-Galactosidase Assays, Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Flow Cytometry, Glycosidase, Neuraminidase, and Related Enzyme Activity, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Permeability | Cell-permeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| Quantity | 5 mg |
| Substrate | Beta-Gal Substrate |
| Substrate Properties | Chemical Substrate |
| Excitation/Emission | 490/514 |
| For Use With (Equipment) | Fluorescence Microscope, Fluorometer, Flow Cytometer |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.