Learn More
Invitrogen™ Fluorescein Di-β-D-Glucuronide (FDGlcU)
Description
Fluorescein di-β-D-glucuronide (FDGlcU) is colorless and nonfluorescent until it is hydrolyzed to the monoglucuronide and then to the highly fluorescent fluorescein.
Beta-Glucuronidase (GUS) Assays, Cell Analysis, Enzyme and Protein Activity Assays, Flow Cytometry, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis, Reporter Assays
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Permeability | Cell-permeant |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Fluorescence |
| Quantity | 5 mg |
| Substrate | Beta-Glucuronidase (GUS) Substrate |
| Substrate Properties | Chemical Substrate |
| Excitation/Emission | 490/514 |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
Unfortunately, we do not have data on the upper limit of solubility for this reagent in PBS. This is something you would need to test empirically.br/>
The molecular weight of the above product is 684.56 g/mol.br/>
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.