Learn More
Invitrogen™ FluoroMyelin™ Red Fluorescent Myelin Stain - Solution in Water
Description
The FluoroMyelin Red fluorescent myelin stain enables quick and selective labeling of myelin in brain cryosections in a single 20-minute labeling step plus washes. This stain can be used in conjunction with antibodies and other dyes, and with standard histochemical methods for cryosection material. Solution in water.
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Red |
| Detection Method | Fluorescence |
| Product Type | Red Fluorescent Myelin Stain |
| Quantity | 1 mL |
| Content And Storage | • Provided as 300X concentration in water. • When stored at 4°C, stable for at least 6 months after receipt. • Protect from light. |
| Label or Dye | FluoroMyelin™ Red |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Fluorescence Microscope |
| Sub Cellular Localization | Myelin |
| Excitation/Emission | 558/654 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.