missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FM™ 1-43 Dye (N-(3-Triethylammoniumpropyl)-4-(4-(Dibutylamino) Styryl) Pyridinium Dibromide)

Product Code. 10769623
Change view
Click to view available options
Quantity:
10 x 100μg
1mg
Unit Size:
1000µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10769623 1mg 1mg
11500786 10 x 100μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10769623 Supplier Invitrogen™ Supplier No. T3163
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Membrane probe

FM 1-43 is an excellent reagent both for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling. This water-soluble dye, which is nontoxic to cells and virtually nonfluorescent in aqueous medium, is believed to insert into the outer leaflet of the cell membrane where it becomes intensely fluorescent. In a neuron that is actively releasing neurotransmitters, the dye becomes internalized within the recycled synaptic vesicles and the nerve terminals become brightly stained. The nonspecific staining of cell-surface membranes can simply be washed off prior to viewing.

FM 1-43 membrane probe is also available as 1mg packaged into 10 vials, each containing 100μg (T-35356). FM 1-43FX membrane probe (F-35355), an analog of FM 1-43 membrane probe that can be fixed in place using aldehyde-based fixatives, is also available.

Cell Analysis, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Endocytosis, Exocytosis & Phagocytosis, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 1mg
Detection Method Fluorescence
Product Line FM
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Plasma Membrane

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.