missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FM™ 4-64FX, fixable analog of FM™ 4-64 membrane stain

Product Code. 11574816
Click to view available options
Quantity:
10 x 100 μg
Unit Size:
1000µg
This item is not returnable. View return policy

Product Code. 11574816

Brand: Invitrogen™ F34653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fixable analog of FM 4-64 membrane stain

The lipophilic probe, FM 4-64FX exhibits low fluorescence in water but fluoresces intensely upon binding the outer leaf of the plasma membrane providing discrete plasma membrane staining. The binding is rapid and reversible, and “FX” denotes that this analog is fixable with aldehyde-based fixatives. It is also an excellent reagent for identifying actively firing neurons and for investigating the mechanisms of activity-dependent vesicle cycling.

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Endocytosis, Exocytosis & Phagocytosis, General Cell Tracing, Microbial Tracking, Neuronal Tracing, Plasma Membrane

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Infrared
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 10 x 100 μg
Detection Method Fluorescence
Product Line FM
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Lipophilic Probe
SubCellular Localization Plasma Membrane

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.