missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HPF (Hydroxyl Radical and Peroxynitrite Sensor)

Product Code. 10134672
Change view
Click to view available options
Quantity:
470 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10134672 470 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10134672 Supplier Invitrogen™ Supplier No. H36004
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Two indicators that together can selectively detect hypochlorite anion

Aminophenyl fluorescein (APF, A36003) and hydroxyphenyl fluorescein (HPF), two reactive oxygen species (ROS) indicators developed by T. Nagano, offer greater specificity and stability than dichlorodihydrofluorescein diacetate (H2DCFDA, D399). APF and HPF show limited non-selective reactivity and relatively high resistance to light-induced oxidation.

  • Fluorescein derivatives are nonfluorescent until they react with hydroxyl radical or peroxynitrite anion
  • APF will also react with hypochlorite anion
  • Two ROS indicators can selectively detect hypochlorite anion
  • Upon oxidation, both APF and HPF exhibit bright green fluorescence (excitation/ emission maxima ∼490/515 nm), making them compatible with any instrument that can detect fluorescein

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 470 μL
Detection Method Fluorescence
Product Type ROS Indicator

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.