missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HybriWell™ Hybridization Sealing System, 40 mm x 21 mm chamber, 0.15 mm deep

Product Code. 10398792
Change view
Click to view available options
Quantity:
1 set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10398792 1 set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10398792 Supplier Invitrogen™ Supplier No. H18210
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Coverslip-seal combination that forms chambers optimized for in situ hybridization work

  • Each disposable incubation chamber seals securely to microscope slide, providing limited-volume chamber for reagent addition
  • Solutions are easily added through dual-access ports without disrupting the specimen
  • Hydrophobic coverslips are nuclease free and will not trap or bind probes, allowing uniform distribution of the reagent over specimen
  • Includes quick-seal tool to secure hydrophobic cover to the microscope slide, as well as nuclease-free adhesive seal tabs to cover the access ports

Cell Analysis, Cellular Imaging, Chromogenic In Situ Hybridization, Fluorescence In Situ Hybridization, In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type Hybridization Sealing System
Product Line HybriWell
Depth (Metric) 0.15 mm
Dimensions 40 x 21 mm
Content And Storage Store at room temperature.
Quantity 1 set
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.