missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Jasplakinolide

Product Code. 11534906
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 11534906

Brand: Invitrogen™ J7473

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Macrocyclic peptide isolated from the marine sponge, Jaspis johnstoni

Jasplakinolide is a potent inducer of actin polymerization in vitro by stimulating actin filament nucleation. Jasplakinolide competes with phalloidin for actin binding (Kd = 15nM).

Cell Analysis, Cell Structure, Cytoskeleton

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
Shipping Condition Room Temperature
Sub Cellular Localization Actin, Cytoskeleton
Molecular Formula C36H45BrN4O6
Physical Form Solid
Product Type Peptide

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.