missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS LipidTOX™ Deep Red Neutral Lipid Stain, for cellular imaging

Product Code. 10242283
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity unitSize
10242283 1 each Each
1 options
This item is not returnable. View return policy

Product Code. 10242283

Brand: Invitrogen™ H34477

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity for neutral lipid droplets

The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/or neutral lipids. HCS LipidTOX Deep Red neutral lipid stain was developed to characterize the potentially toxic effects of compounds on lipid metabolism in mammalian cell lines. The LipidTOX neutral lipid stain has an extremely high affinity for neutral lipid droplets and can be detected by fluorescence microscopy or an HCS reader. This probe is compatible with the HCS LipidTOX phospholipidosis detection reagents. HCS LipidTOX neutral lipid stains can also be used to monitor the formation and differentiation of adipocytes, a process called adipogenesis.

Cell Analysis, Cellular Imaging, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Deep Red
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) High Content Instrument
Quantity 1 each
Detection Method Fluorescence
Product Line LipidTOX
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Lipid Stain
SubCellular Localization Cell Membranes, Lipids

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.