Learn More
Invitrogen™ HCS LipidTOX™ Green Neutral Lipid Stain, for cellular imaging
Beskrivelse
The intracellular accumulation of neutral lipids, steatosis, is often triggered by drugs that affect the metabolism of fatty acids and/or neutral lipids. HCS LipidTOX Green neutral lipid stain was developed to characterize the potentially toxic effects of compounds on lipid metabolism in mammalian cell lines. The LipidTOX neutral lipid stain has an extremely high affinity for neutral lipid droplets and can be detected by fluorescence microscopy or an HCS reader. This probe is compatible with the HCS LipidTOX phospholipidosis detection reagents. HCS LipidTOX neutral lipid stains can also be used to monitor the formation and differentiation of adipocytes, a process called adipogenesis.
Cell Analysis, Cellular Imaging, High-Content Screening (HCS)
Ordreinfo
Shipping Condition: Room temperature
Tekniske data
Tekniske data
| Color | Green |
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| For Use With (Equipment) | High Content Instrument |
| Quantity | 1 each |
| Detection Method | Fluorescence |
| Product Line | LipidTOX |
| Shipping Condition | Room Temperature |
| Label Type | Fluorescent Dye |
| Product Type | Lipid Stain |
| Sub Cellular Localization | Cell Membranes, Lipids |
For Research Use Only. Not for use in diagnostic procedures.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.