missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lucifer Yellow CH, Potassium Salt

Product Code. 11524956
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11524956 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11524956 Supplier Invitrogen™ Supplier No. L1177
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Water-soluble dye with excitation/emission peaks of 428/536nm

  • Tool for studying neuronal morphology, because it contains carbohydrazide (CH) group that allows it to be covalently linked to surrounding biomolecules during aldehyde fixation

Cell Analysis, Cell Tracing and Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Detection Method Fluorescence
Product Type Potassium Salt
Quantity 25 mg
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Cytoplasm & Cytosol
Excitation/Emission 428/536 nm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.