missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LysoTracker™ Blue DND-22, special packaging

Product Code. 12080146
Click to view available options
Quantity:
20 x 50 μL
Unit Size:
1000µL
This item is not returnable. View return policy

Product Code. 12080146

Brand: Invitrogen™ L7525

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Special packaging

LysoTracker Blue DND-22 is a cell-permeable, non-fixable, blue fluorescent dye that stains acidic compartments within live cells, such as lysosomes. Blue DND-22 has an excitation and emission maximum of 373/422 nm and can be efficiently excited using a DAPI filter. LysoTracker probes are available in a variety of fluorescent colors.

Simple, highly specific, one-step staining and tracking of acidic organelles
LysoTracker probes consist of a hydrophobic fluorophore linked to a weak base that is only partially protonated at neutral pH. In a neutrally charge state, LysoTracker probes can freely diffuse across intact plasma membranes of live cells. Due to the inherently acidic properties within the lysosome, upon diffusion into the lysosome the weakly basic moiety is protonated. In this charged state, the probe does not readily diffuse across the organelle membrane, providing a localized accumulation for distinct staining of acidic organelles such as lysosomes. Effective at nanomolar concentrations, LysoTracker probes are highly selective for acidic organelles and provide simple one-step staining that does not rely on antibody detection.

Lysosomes are acidic in nature due to the production and storage of digestive enzymes (hydrolases). The acidic environment of lysosomes enables degradation of carbohydrates, lipids, nucleic acids, and peptides. Extracellular proteins, virus or bacteria can also be internalized and trafficked to the lysosome for degradation via the lysosomal proteolysis pathway. In addition, misfolded proteins or damaged organelles become targets for lysosomal degradation via the induction of autophagy. Autophagy is important for cellular differentiation, survival during nutrient deprivation, and normal growth control. The inherent acidic nature within lysosomes, and any change of pH during biosynthesis or pathogenesis, can be exploited by various probes that respond to an acidic environment. LysoTracker products provide fluorescence detection for live-cell staining of acidic environments and can be used for labeling and tracing acidic organelles such as lysosomes.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Concentration 1 mM
Content And Storage Store desiccated at ≤–20°C
• Protect from light
• Avoid freeze-thaw cycles and do not store in a frost-free freezer
• Store in single-use aliquots, if possible

Excitation Wavelength Range 373/422
For Use With (Equipment) Fluorescence Microscope, Fluorescent Imager
Quantity 20 x 50 μL
Detection Method Fluorescence
Description LysoTracker™ Blue DND-22
Emission Blue
Form Liquid
Product Line LysoTracker
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
SubCellular Localization Lysosomes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.