missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LysoTracker™ Green DND-26, special packaging

Product Code. 11594976
Change view
Click to view available options
Quantity:
20 x 50 μL
Unit Size:
1000µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11594976 20 x 50 μL 1000µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11594976 Supplier Invitrogen™ Supplier No. L7526
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Special packaging

LysoTracker Green DND-26 is a cell-permeable, non-fixable, green fluorescent dye that stains acidic compartments within a cell, such as lysosomes. Green DND-26 has an excitation and emission maximum of 504/511 nm and can be efficiently excited using a FITC filter. LysoTracker probes are available in a variety of fluorescent colors.

Simple, highly specific, one-step staining and tracking of acidic organelles
LysoTracker probes consist of a hydrophobic fluorophore linked to a weak base that is only partially protonated at neutral pH. In a neutrally charge state, LysoTracker probes can freely diffuse across intact plasma membranes of live cells. Due to the inherently acidic properties within the lysosome, upon diffusion into the lysosome the weakly basic moiety is protonated. In this charged state, the probe does not readily diffuse across the organelle membrane, providing a localized accumulation for distinct staining of acidic organelles such as lysosomes. Effective at nanomolar concentrations, LysoTracker probes are highly selective for acidic organelles and provide simple one-step staining that does not rely on antibody detection.

Lysosomes are acidic in nature due to the production and storage of digestive enzymes (hydrolases). The acidic environment of lysosomes enables degradation of carbohydrates, lipids, nucleic acids, and peptides. Extracellular proteins, virus or bacteria can also be internalized and trafficked to the lysosome for degradation via the lysosomal proteolysis pathway. In addition, misfolded proteins or damaged organelles become targets for lysosomal degradation via the induction of autophagy. Autophagy is important for cellular differentiation, survival during nutrient deprivation, and normal growth control. The inherent acidic nature within lysosomes, and any change of pH during biosynthesis or pathogenesis, can be exploited by various probes that respond to an acidic environment. LysoTracker products provide fluorescence detection for live-cell staining of acidic environments and can be used for labeling and tracing acidic organelles such as lysosomes.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Concentration 1 mM
Content And Storage Store desiccated at ≤–20°C
• Protect from light
• Avoid freeze-thaw cycles and do not store in a frost-free freezer
• Store in single-use aliquots, if possible

Excitation Wavelength Range 504/511
For Use With (Equipment) Fluorescence Microscope, Fluorescent Imager
Quantity 20 x 50 μL
Detection Method Fluorescence
Description LysoTracker™ Green DND-26
Emission Green
Product Line LysoTracker
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Lysosomes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.