missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MANT-ADP (2'-(or-3')-O-(N-Methylanthraniloyl) Adenosine 5'-Diphosphate, Disodium Salt)

Product Code. 10174812
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10174812 400 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10174812 Supplier Invitrogen™ Supplier No. M12416
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produces nucleotide analogs causing little perturbation of nucleotide-protein interactions

The nucleotide analog MANT ADP is modified on the ribose moiety. The compact mature of the MANT fluorophore and its attachment position results in nucleotide analogs that induce minimal perturbation of nucleotide-protein interactions. Because MANT fluorescence is sensitive to the environment of the fluorophore, nucleotide-protein interactions may be directly detectable. MANT nucleotides are valuable probes of the structure and enzymatic activity of nucleotide-binding proteins.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type MANT-ADP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye MANT (N-Methylanthraniloyl)
Quantity 400 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.