missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUP (4-Methylumbelliferyl Phosphate, Free Acid)

Product Code. 10013142
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10013142 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10013142 Supplier Invitrogen™ Supplier No. M6491
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MUP is a fluorescent substrate for alkaline phosphatases

Alkaline phosphatase mediated hydrolysis of its phosphate substituent yields the blue-fluorescent 4-methylumbelliferyl (excitation/emission approx. 386/448nm).

Alkaline Phosphatase Assays, Biochemical Toxicology Assays, Cell Analysis, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Reporter Assays, Target and Lead Identification and Validation, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Powder
Quantity 1 g
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Alkaline Phosphatase
Excitation/Emission 360⁄449
For Use With (Equipment) Fluorometer, Microplate Reader
Label or Dye MUP (4-methylumbelliferyl phosphate)
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.