missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBD Cholesterol (22-(N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)Amino)-23,24-Bisnor-5-Cholen-3β-Ol)

Product Code. 11559116
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11559116 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11559116 Supplier Invitrogen™ Supplier No. N1148
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

This fluorescent analog, NBD cholesterol, is an environment-sensitive probe that localizes in the membrane's interior and is useful for investigating lipid transport processes as well lipid-protein interactions.

This fluorescent analog, NBD cholesterol, is an environment-sensitive probe that localizes in the membrane's interior and is useful for investigating lipid transport processes as well lipid-protein interactions.

We recommend dissolving in chloroform (CHCl3) or acetonitrile (MeCN) to attain a stock concentration up to 1 mM.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cholesterol Analogs
Quantity 10 mg
Physical Form Solid
Product Line Invitrogen
Excitation/Emission 469/537 nm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.