missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nigericin, Free Acid

Product Code. 11589116
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11589116 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11589116 Supplier Invitrogen™ Supplier No. N1495
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

For intracellular calibration of fluorescence response of cytosolic pH indicators

  • Performed using K+/H+ ionophore nigericin
  • It causes equilibration of intracellular and extracellular pH inpresence of depolarizing concentration of extracellular K+

Anion Detection, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Shipping Condition Room Temperature
Indicator Ionophore
Molecular Formula C40H68O11
Physical Form Solid
Product Type Potassium Ionophore

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.