missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPE-caged ATP (Adenosine 5'-Triphosphate, P3-(1-(2-Nitrophenyl)Ethyl) Ester, Disodium Salt)

Product Code. 11529046
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11529046 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11529046 Supplier Invitrogen™ Supplier No. A1048
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Flash UV photolysis of caging group creates release of free nucleotides

NPE-caged ATP is a nucleotide analog in which the terminal phosphate is esterified with a blocking group, rendering the molecule biologically inactive. Flash photolysis of the caging group with UV illumination at 360nm results in a rapid and highly localized release of the free nucleotide at the site of illumination.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type NPE-Caged ATP
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Shipping Condition Wet Ice
Label or Dye NPE (P3-(1-(2-nitrophenyl)ethyl) ester)
Quantity 5 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.