missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gelatin From Pig Skin, Oregon Green™ 488 Conjugate

Product Code. 11594856
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11594856 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11594856 Supplier Invitrogen™ Supplier No. G13186

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Can be used for investigation of gelatinases and collagenases

Molecular Probes fluorescent conjugates of gelatin are designed for researchers studying not only collagen-binding proteins and collagen metabolism, but also for the study of gelatinases and collagenases (metalloproteins that digest collagen and gelatin).

  • Label (Ex/Em): Oregon Green™ 488 (approx. 499/519nm)
  • Degree of labeling is listed on the product label (based on molecular weight of 100,000)
  • Two fluorescent conjugates of gelatin are available—one in which gelatin is coupled to fluorescein and the other in which gelatin is coupled to Oregon Green 488 dye.

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Oregon Green 488
Species Pig
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Classic Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Gelatin
Product Line Oregon Green

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.