missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Paclitaxel (Taxol Equivalent) - for use in research only

Product Code. 11579296
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11579296 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11579296 Supplier Invitrogen™ Supplier No. P3456
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

For use in research only

Paclitaxel (formerly referred to as taxol in some scientific literature) is the approved generic name for the anticancer pharmaceutical Taxol (a registered trademark of Bristol-Myers Squibb Co). Paclitaxel promotes tubulin assembly, producing aggregates that cannot be depolymerized by dilution, calcium ions, cold or a number of microtubule-disrupting drugs. Cultured cells treated with paclitaxel are blocked in the G2 and M phases of the cell cycle.

Cell Analysis, Cell Structure, Cytoskeleton

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Quantity 5 mg
Form Lyophilized
Inhibitors Cytoskeletal Inhibitor
Shipping Condition Room Temperature
Specific Inhibit or Target Tubulin
Product Type Paclitaxel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.