missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PeppermintStick™ Phosphoprotein Molecular Weight Standards

Product Code. 10267432
Change view
Click to view available options
Quantity:
40 μL
400 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10267432 40 μL Each
10358242 400 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10267432 Supplier Invitrogen™ Supplier No. P27167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins ranging from 14.4 to 116 kDa.

PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins ranging from 14.4 to 116 kDa. Separation by polyacrylamide gel electrophoresis resolves the standard mixture into two phosphorylated and four nonphosphorylated protein bands.

Applications

  • Accurate protein sizing on SDS-polyacrylamide gels and western blots.
  • Positive and negative controls for detection of phosphorylated proteins, such as the methods provided by our Pro-Q™ Diamond phosphoprotein gel stain

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 40 μL
Stain Type Fluorescent

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.