missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Resorufin β-D-Galactopyranoside

Product Code. 11513677
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11513677 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11513677 Supplier Invitrogen™ Supplier No. R1159
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For the hydrolysis product, resorufin

The fluorogenic β-galactosidase substrate resorufin β-D-galactopyranoside yields the hydrolysis product, resorufin, with absorption/emission maxima at 571/585nm.

Beta-Galactosidase Assays, Cell Analysis, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Quantity 25 mg
Substrate Beta-Gal Substrate
Substrate Properties Chemical Substrate
Excitation/Emission 571/585
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.