missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Secure-Seal™ Spacer, one well, 20 mm diameter, 0.12 mm deep

Product Code. 10206932
Change view
Click to view available options
Quantity:
1 set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10206932 1 set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206932 Supplier Invitrogen™ Supplier No. S24736
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Removable hydrophobic barrier that provides flexibility for isolating specimens

  • Barriers may be used to isolate cells grown in culture dishes or to separate specimens on microscopy slides during staining procedures
  • Silicone material is autoclavable and adheres to any smooth surface

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type Spacer
Product Line Secure-Seal
Depth (Metric) 0.12 mm
Diameter (Metric) 20 mm
Content And Storage Store at room temperature.
Quantity 1 set
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.