missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Singlet Oxygen Sensor Green - Special Packaging

Product Code. 11580676
Change view
Click to view available options
Quantity:
10 x 100 μg
Unit Size:
1000µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11580676 10 x 100 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11580676 Supplier Invitrogen™ Supplier No. S36002
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detection reagent highly selective for singlet oxygen

Singlet Oxygen Sensor Green is a detection reagent that is highly selective for singlet oxygen. Unlike other available fluorescent and chemiluminescent singlet oxygen detection reagents, Singlet Oxygen Sensor Green does not show any appreciable response to hydroxyl radical or superoxide. This indicator initially exhibits weak blue fluorescence, but in the presence of singlet oxygen, it emits a green fluorescence (excitation/emission maxima ∼504/525 nm) similar to that of fluorescein. Singlet Oxygen Sensor Green is cell-impermeant.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 100 μg
For Use With (Application) ROS
Detection Method Fluorescence
Product Type ROS Indicator
Excitation Wavelength Range 504 nm
Emission Wavelength Range 525 nm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.