Learn More
Invitrogen™ SYTO™ 41 Blue Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Description
The cell-permeant SYTO 41 blue fluorescent nucleic acid stain exhibits bright, blue fluorescence upon binding to nucleic acids. Because the staining pattern of the SYTO dyes in live cells may vary from one cell type to another, we offer the SYTO Blue Fluorescent Nucleic Acid Stain Sampler Kit (Cat. No. S-11350) to enable researchers to find the dye that works best for a particular application.
Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Blue |
| Content And Storage | Store in freezer at -5°C to -30°C and protect from light. |
| Excitation Wavelength Range | 430 nm |
| Dye Type | Cell-Permeant |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 250 μL |
| Volume (Metric) | 250 μL |
| Detection Method | Fluorescence |
| Description | SYTO™ 41 Blue Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO |
| Emission | 454 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.