missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tali™ Apoptosis Kit - Annexin V Alexa Fluor™ 488 & Propidium Iodide

Product Code. 11370942
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370942 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11370942 Supplier Invitrogen™ Supplier No. A10788
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Annexin V – Alexa Fluor 488 conjugate, Annexin Binding solution, Propidium Iodide (PI) solution

Discrimination of apoptotic from necrotic and live cells in a population

The TaliApoptosis Kit enables identification of apoptotic cells. The kit stains apoptotic cells with green Annexin V – Alexa Fluor™ 488, stains necrotic cells with both red propidium iodide and green Annexin V – Alexa Fluor 488, and does not stain live cells.

Apoptosis is a carefully regulated process of cell death that occurs as a normal part of development. Inappropriately regulated apoptosis is implicated in disease states, such as Alzheimer's disease and cancer. Apoptosis is distinguished from necrosis by characteristic morphological and biochemical changes, including compaction and fragmentation of the nuclear chromatin, shrinkage of the cytoplasm, and loss of membrane asymmetry.

Easily Discriminate Apoptotic from Necrotic Cells with Two-color Staining
In normal live cells, phosphatidyl serine (PS) is located on the cytoplasmic surface of the cell membrane. In apoptotic cells, however, PS is translocated from the inner to the outer surface of the plasma membrane, exposing it to the extracellular environment. The human anticoagulant Annexin V displays a high affinity for PS. Annexin V labeled with a fluorophore such as Alexa Fluor 488 can be used to identify apoptotic cells by binding to the exposed PS.

The TaliApoptosis Kit includes a solution of propidium Iodide (PI). PI is a cell-impermeant fluorogenic DNA-binding dye used for years to identify necrotic cells. PI is impermeant to live cells, but easily enters dead cells where it binds to nucleic acids and becomes fluorescent.

After staining a cell population using the TaliApoptosis Assay Kit with Annexin V-Alexa Fluor 488 and Propidium Iodide, apoptotic cells display green fluorescence, dead cells display red and green fluorescence and live cells show little or no fluorescence.

Annexin V Translocation, Apoptosis, Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability, Proliferation and Function, Cellular Imaging, Imaging Cytometry

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format Vial
Product Type Apoptosis Kit
Content And Storage Tali™ - Annexin V Alexa Fluor™: 488: Store 2 – 6°C
Tali™ - Propidium Iodide: Store 2 – 6°C
Tali™ - Annexin V Binding Buffer: Store 2 – 6°C
Shipping Condition Wet Ice
Product Line Tali
Conjugate Alexa Fluor 488
For Use With (Equipment) Tali™ Image-Based Cytometer
What are the fluorescence excitation/emission maxima for the dyes contained in the Tali Apoptosis Kit - Annexin V Alexa Fluor 488 & Propidium Iodide?

Annexin V Alexa Fluor 488: 488/499 nm
Propidium Iodide: 535/617 nm, bound to DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.