missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Albumin from Bovine Serum (BSA), Texas Red™ conjugate
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
- Applications include studying rate of endocytosis and exocytosis and following intracellular protein processing
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Quantity | 5 mg |
| Conjugate | Texas Red |
| Species | Bovine |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Classic Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Albumin Proteins |
| Product Line | Texas Red™ |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction