missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thapsigargin

Product Code. 10226822
Change view
Click to view available options
Quantity:
1 mg
20 x 50 μg
Unit Size:
1000µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10226822 1 mg 1mg
11570796 20 x 50 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10226822 Supplier Invitrogen™ Supplier No. T7458
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Tumor promoter

Thapsigargin is a naturally occurring sesquiterpene lactone isolated from the umbelliferous plant Thapsia garganica. This tumor promoter releases Ca2++ from intracellular stores by specifically inhibiting the endoplasmic reticulum Ca2++-ATPase. It does not directly affect plasma membrane Ca2++-ATPases, Ins 1,4,5-P3 production or protein kinase C activity.

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Label Type Unlabeled
Molecular Formula C34H50O12
Physical Form Solid
Product Type Inhibitor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.