missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thapsigargin

Product Code. p-4921138
Click to view available options
Quantity:
1 mg
20 x 50 μg
Unit Size:
1000µg
1mg
This item is not returnable. View return policy

Product Code. 10226822

Brand: Invitrogen™ T7458

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Tumor promoter

Thapsigargin is a naturally occurring sesquiterpene lactone isolated from the umbelliferous plant Thapsia garganica. This tumor promoter releases Ca2++ from intracellular stores by specifically inhibiting the endoplasmic reticulum Ca2++-ATPase. It does not directly affect plasma membrane Ca2++-ATPases, Ins 1,4,5-P3 production or protein kinase C activity.

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Receptors

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Shipping Condition Room Temperature
Label Type Unlabeled
Molecular Formula C34H50O12
Physical Form Solid
Product Type Inhibitor

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.