missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED™, J.T. Baker™

Product Code. 10152500 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10152500 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10152500 Supplier JT Baker Supplier No. 0810.1000
  • ONLINE EXCLUSIVE PRICE
    314.00 €
    203.01 € / 1kg
    Save 110.99 €
    35% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Density 1.1 - 2.2 g/cm3 (20°C)
Quantity 1 kg
CAS 70955-01-0
Molecular Formula C20H25FN2O8
MDL Number MFCD00131613
Synonym Linde 4 A
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
IUPAC Name Molecular sieves
Molecular Weight (g/mol) 440.42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.